missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apilimod, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Apilimod (STA 5326) is a potent IL-12/IL-23 inhibitor, and strongly inhibits IL-12 with IC50s of 1 nM and 2 nM, in IFN-γ/SAC-stimulated human PBMCs and SAC-treated monkey PBMCs, respectively. Apilimod is a potent and highly selective PIKfyve inhibitor.
Specifications
Specifications
| Chemical Name or Material | Apilimod |
| Physical Form | Solid |
| CAS | 541550-19-0 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C23H26N6O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | STA 5326 |
| Solubility Information | DMSO : 100 mg/mL (238.95 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction