missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Apilimod dimesylate
GREENER_CHOICE

Product Code. p-7235827
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18767466

Brand: Tocris Bioscience™ 7283/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PIKfyve inhibitor

Apilimod dimesylate is a potent and selective PIKfyve inhibitor (IC50 = 14 nM). Exhibits no significant activity at other lipid kinases and protein kinases, including PIP4K, PIP5K, mTOR, PI3K and PI4K. Inhibits cellular entry by SARS-CoV-2, MERS-CoV and MHV S pseudovirions. Reduces number of SARS-CoV-2-infected hiPSC-derived pneumocyte-like cells by 85% and inhibits SARS-CoV-2 replication in donor lung tissue. Also exhibits selective cytotoxicity in B-cell non-Hodgkin lymphoma compared with normal cells. Inhibits production of IL-12 and IL-23. Anti-inflammatory. Water-soluble.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(6-(2-(3-Methylbenzylidene)hydrazinyl)-2-(2-(pyridin-2-yl)ethoxy)pyrimidin-4-yl)morpholine dimethanesulfonate
CAS 870087-36-8
Quantity 25 mg
Target PIKFyve/PIP5K3
Molecular Formula C23H26N6O20.2 CH3SO3H
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.