missing translation for 'onlineSavingsMsg'
Learn More
Learn More
API5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46223
This item is not returnable.
View return policy
Description
API5 Polyclonal specifically detects API5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| API5 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| AAC-11API5L1, AAC11fibroblast growth factor 2-interacting factor 2, Antiapoptosis clone 11 protein, API-5, apoptosis inhibitor 5, Cell migration-inducing gene 8 protein, Fibroblast growth factor 2-interacting factor, FIF, migration-inducing protein MIG8, Protein XAGL | |
| Rabbit | |
| 58 kDa | |
| 0.1 mL | |
| Apoptosis | |
| 8539 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin | |
| Q9BZZ5 | |
| API5 | |
| The immunogen for this product maps to a region between residue 1 and 50 of human Apoptosis Inhibitor 5 using the numbering given in entry XP_508375.2 (GeneID 8539). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction