missing translation for 'onlineSavingsMsg'
Learn More

API-1, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18015342
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015342 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18015342 Supplier Tocris Bioscience Supplier No. 3897/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective Akt/PKB inhibitor. Antitumor

Akt/protein kinase B (PKB) inhibitor. Binds the pleckstrin homology domain of Akt and blocks Akt membrane translocation. Inhibits EGF-induced kinase activity of Akt1, Akt2 and Akt3. Induces cell growth arrest and apoptosis in human cancer cells expressing constitutively active Akt. Displays antitumor activity in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 36707-00-3
Molecular Formula C13H15N5O6
Quantity 10 mg
Synonym api-1, 4-amino-8-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-5-oxo, 4-amino-5,8-dihydro-5-oxo-8-?-d-ribofuranosyl-pyrido 2,3-d pyrimidine-6-carboxamide, 4-amino-5,8-dihydro-5-oxo-8-beta-d-ribofuranosyl-pyrido 2,3-d pyrimidine-6-carboxamide, 4-amino-8-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-5-oxopyrido 2,3-d pyrimidine-6-carboxamide
InChI Key SPBWHPXCWJLQRU-FITJORAGSA-N
SMILES C1=C(C(=O)C2=C(N1C3C(C(C(O3)CO)O)O)N=CN=C2N)C(=O)N
IUPAC Name 4-amino-8-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-oxopyrido[2,3-d]pyrimidine-6-carboxamide
Molecular Weight (g/mol) 337.292
PubChem CID 24773090
Formula Weight 337.29
Percent Purity >96%
Chemical Name or Material API-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.