missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523466
This item is not returnable.
View return policy
Description
The amino acid sequence used as immunogen is 88% homologous in Chicken.
APH1 is an essential subunit of the gamma-secretase complex, an endoprotease complex that catalyzes the intramembrane cleavage of integral proteins such as Notch receptors and APP (beta-amyloid precursor protein). The gamma-secretase complex is minimally composed of a presenilin homodimer (PSEN1 or PSEN2), nicastrin (NCSTN), APH1 (APH1A or APH1B) and PEN2, although other components may exist. APH1 probably represents a stabilizing cofactor for the presenilin homodimer that promotes the formation of a stable complex.
Specifications
| APH1 | |
| Polyclonal | |
| Unconjugated | |
| APH1A | |
| 6530402N02Rik; anterior pharynx defective 1 homolog A; anterior pharynx defective 1a homolog; anterior pharynx defective 1a homolog (C. elegans); APH1; APH-1; aph-1 homolog A, gamma secretase subunit; aph-1 homolog A, gamma-secretase subunit; Aph1a; APH-1a; APH1A gamma secretase subunit; Aph-1alpha; CGI-78; Gamma-secretase subunit APH-1A; hypothetical protein LOC539247; presenilin-stabilization factor; PSF; RGD1311546; RP4-790G17.3; UNQ579/PRO1141 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100054581, 100855839, 51107, 539247 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Q96BI3 | |
| APH1A | |
| A portion of amino acids 80-145 of human APH1A. | |
| 100 μg | |
| Primary | |
| Human, Canine, Monkey, Bovine, Horse, Marsupial | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction