missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APH1 Polyclonal Antibody

Product Code. 12858502
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12858502 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12858502 Supplier Invitrogen™ Supplier No. PA520318

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Despite its predicted molecular weight, APH1 often migrates at aberrant locations in SDS-PAGE. A suggested positive control is human brain tissue lysate. PA5-20318 can be used with blocking peptide PEP-0438.

APH1 is an essential subunit of the gamma-secretase complex, an endoprotease complex that catalyzes the intramembrane cleavage of integral proteins such as Notch receptors and APP (beta-amyloid precursor protein). The gamma-secretase complex is minimally composed of a presenilin homodimer (PSEN1 or PSEN2), nicastrin (NCSTN), APH1 (APH1A or APH1B) and PEN2, although other components may exist. APH1 probably represents a stabilizing cofactor for the presenilin homodimer that promotes the formation of a stable complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APH1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene APH1A
Gene Accession No. Q8BVF7, Q96BI3
Gene Alias 6530402N02Rik; anterior pharynx defective 1 homolog A; anterior pharynx defective 1a homolog; anterior pharynx defective 1a homolog (C. elegans); APH1; APH-1; aph-1 homolog A, gamma secretase subunit; aph-1 homolog A, gamma-secretase subunit; Aph1a; APH-1a; APH1A gamma secretase subunit; Aph-1alpha; CGI-78; Gamma-secretase subunit APH-1A; hypothetical protein LOC539247; presenilin-stabilization factor; PSF; RGD1311546; RP4-790G17.3; UNQ579/PRO1141
Gene Symbols APH1A
Host Species Rabbit
Immunogen An 18 amino acid peptide from near the center of human APH1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226548, 365872, 51107
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.