missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APG8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-471-0.25MG
This item is not returnable.
View return policy
Description
APG8 Polyclonal specifically detects APG8 in Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| APG8 | |
| Polyclonal | |
| Western Blot 1:4000-1:8000, ELISA 1:20000-1:100000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| APG8, Atg8p, AUT7, CVT5 | |
| Rabbit | |
| 0.25 mg | |
| Atg 8 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| Atg8p | |
| This purified antibody was prepared from rabbit serum after repeated immunizations with recombinant yeast APG8 protein. | |
| 852200 | |
| Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction