missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APEX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572607
This item is not returnable.
View return policy
Description
APEX2 Polyclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot
Apurinic/apyrimidinic (AP) sites occur frequently in DNA molecules by spontaneous hydrolysis, by DNA damaging agents or by DNA glycosylases that remove specific abnormal bases. AP sites are pre-mutagenic lesions that can prevent normal DNA replication so the cell contains systems to identify and repair such sites. Class II AP endonucleases cleave the phosphodiester backbone 5' to the AP site. This gene encodes a protein shown to have a weak class II AP endonuclease activity. Most of the encoded protein is located in the nucleus but some is also present in mitochondria. This protein may play an important role in both nuclear and mitochondrial base excision repair (BER).
Specifications
| APEX2 | |
| Polyclonal | |
| Unconjugated | |
| APEX2 | |
| AP endonuclease 2; AP endonuclease XTH2; Ape2; APEX nuclease (apurinic/apyrimidinic endonuclease) 2; APEX nuclease 2; APEX nuclease-like 2; APEX2; APEXL2; apurinic/apyrimidinic endodeoxyribonuclease 2; apurinic/apyrimidinic endonuclease 2; apurinic/apyrimidinic endonuclease-like 2; Apurinic-apyrimidinic endonuclease 2; C430040P13Rik; DNA-(apurinic or apyrimidinic site) lyase 2; RP5-884M20.3; XTH2; ZGRF2; zinc finger, GRF-type containing 2 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 27301 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9UBZ4 | |
| APEX2 | |
| KLH conjugated synthetic peptide between 143-171 amino acids from the Central region of human APEX2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction