missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APEX2 Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue, HEK293 whole cell, COS-7 whole cell. Flow: A549 cell.
Apurinic/apyrimidinic (AP) sites occur frequently in DNA molecules by spontaneous hydrolysis, by DNA damaging agents or by DNA glycosylases that remove specific abnormal bases. AP sites are pre-mutagenic lesions that can prevent normal DNA replication so the cell contains systems to identify and repair such sites. Class II AP endonucleases cleave the phosphodiester backbone 5' to the AP site. This gene encodes a protein shown to have a weak class II AP endonuclease activity. Most of the encoded protein is located in the nucleus but some is also present in mitochondria. This protein may play an important role in both nuclear and mitochondrial base excision repair (BER).
Specifications
Specifications
| Antigen | APEX2 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and 0.05mg sodium azide |
| Gene | APEX2 |
| Gene Accession No. | Q68G58, Q9UBZ4 |
| Gene Alias | AP endonuclease 2; AP endonuclease XTH2; Ape2; APEX nuclease (apurinic/apyrimidinic endonuclease) 2; APEX nuclease 2; APEX nuclease-like 2; APEX2; APEXL2; apurinic/apyrimidinic endodeoxyribonuclease 2; apurinic/apyrimidinic endonuclease 2; apurinic/apyrimidinic endonuclease-like 2; Apurinic-apyrimidinic endonuclease 2; C430040P13Rik; DNA-(apurinic or apyrimidinic site) lyase 2; RP5-884M20.3; XTH2; ZGRF2; zinc finger, GRF-type containing 2 |
| Gene Symbols | APEX2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?