missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APEX2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92295-0.02ml
This item is not returnable.
View return policy
Description
APEX2 Polyclonal antibody specifically detects APEX2 in Human samples. It is validated for Western Blot
Specifications
| APEX2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| AP endonuclease 2, APE2, APEX nuclease (apurinic/apyrimidinic endonuclease) 2, APEX nuclease 2, APEXL2, Apurinic-apyrimidinic endonuclease 2, DNA-(apurinic or apyrimidinic site) lyase 2, DNA-apurinic or apyrimidinic site lyase 2, EC 3.1, EC 4.2.99.18, XTH2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human APEX2 (NP_055296.2). MLRVVSWNINGIRRPLQGVANQEPSNCAAVAVGRILDELDADIVCLQETKVTRDALTEPLAIVEGYNSYFSFSRNRSGYSGVATFCKDNATPVAAEEGLSGLFATQNGDVGCYGNMDEFTQEELRALDSEGRALLTQHKIRTWEGKEKTLTLINVYCPHADPGRPERLVFKMRFYRLLQI | |
| 0.02 mL | |
| Direct Reversal of DNA Damage | |
| 27301 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction