missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ APET Liners for Safety Trays

Environmental friendly, easy to clean and maintain for routine work and storage in safe areas.

Brand:  Fisherbrand™

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12327207

  • £119.00 / Pack of 25
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

  • Clears up spillages easily
  • Environmental friendly
Specifications

Specifications

540 x 570mm
APET
570 mm
25
540 mm
Liners
Radiation, Biohazard, General purpose Trays
Rectangular
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers