missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apelin Receptor Recombinant Rabbit Monoclonal Antibody (HL2980)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556597
This item is not returnable.
View return policy
Description
Apelin Receptor Recombinant Monoclonal Antibody for IHC (P)
APLNR (Apelin Receptor) is a member of the G protein-coupled receptor gene family. The encoded protein is related to the angiotensin receptor, but is actually an apelin receptor that inhibits adenylate cyclase activity and plays a counter-regulatory role against the pressure action of angiotensin II by exerting hypertensive effect. It functions in the cardiovascular and central nervous systems, in glucose metabolism, in embryonic and tumor angiogenesis and as a human immunodeficiency virus (HIV-1) coreceptor. Two transcript variants resulting from alternative splicing have been identified.
Specifications
| Apelin Receptor | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P35414 | |
| Aplnr | |
| Recombinant protein encompassing a sequence within the Intracellular domain of human Apelin Receptor. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| HL2980 | |
| Unconjugated | |
| Aplnr | |
| AGTRL1; angiotensin II receptor-like 1; angiotensin receptor-like 1; Apelin receptor; APJ; APJ (apelin) receptor; APJ receptor; APJR; Aplnr; B78; G protein-coupled receptor APJ; GPCR34; G-protein coupled receptor APJ; G-protein coupled receptor HG11; HG11; HG11 orphan receptor; MSR; msr/apj | |
| Rabbit | |
| Protein A | |
| RUO | |
| 187 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction