missing translation for 'onlineSavingsMsg'
Learn More

Apelin Antibody (1A8-2A5), Novus Biologicals™

Product Code. 18368938 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368938 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368938 Supplier Novus Biologicals Supplier No. H00008862M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Apelin Monoclonal antibody specifically detects Apelin in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifica

Antigen Apelin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1A8-2A5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH21104.1
Gene Alias AGTRL1 ligand, APEL, apelin, apelin, AGTRL1 ligand, APJ endogenous ligand, XNPEP2
Host Species Mouse
Immunogen APLN (AAH21104.1, 1 a.a. ~ 122 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSATWCSPEGQGMGQGPGREVGGNSAASGPASPIRDPCLSEAGLKGPPSAHPRRLCLLHRLVCFSGGLTSIQLSPRTCCSHQWAQLFSPACFPQWRAPGCSLDDSRSLTRIRPVHLPGPSLD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8862
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.