Learn More
DiscoverX Apelin-13 TFA salt
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Apelin-13 is a bioactive neuropeptide processed from the pre-proprotein apelin that is active in the nervous system. It is an endogenous ligand of the receptor APJ. Plays a role in cardiovascular regulation, fluid homeostasis, hypertension, and insulin sensitivity. May undergo pyroglutamylation, and may block HIV-1 and HIV-2 entry into cells expressing the APJ receptor. Compared to apelin-36, blocks HIV entry poorly. Provided as 1mg of dry powder alongside reconstitution buffer.
Specifications
Specifications
| Product Type | Control Ligands |
| Quantity | 1 mg |