missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APEG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99997-100ul
This item is not returnable.
View return policy
Description
APEG1 Polyclonal antibody specifically detects APEG1 in Human samples. It is validated for ELISA
Specifications
| APEG1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| aortic preferentially expressed gene 1, Aortic preferentially expressed protein 1, APEG1, BPEG, EC 2.7.11, EC 2.7.11.1, KIAA1297APEG-1, MGC12676, nuclear protein, marker for differentiated aortic smooth muscle anddown-regulated with vascular injury, SPEG complex locus, SPEGalpha, SPEGbeta, striated muscle preferentially expressed protein kinase | |
| Produced in rabbits immunized with purified, recombinant Human APEG1 (Uniprot#: Q15772-4; Met1-Glu113). | |
| 100 μg | |
| Cell Cycle and Replication | |
| 10290 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction