missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APE Rabbit anti-Human, Mouse, Rat, Clone: S07-5A9, Novus Biologicals™
Rabbit Monoclonal Antibody
£222.00 - £451.00
Specifications
| Antigen | APE |
|---|---|
| Clone | S07-5A9 |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunofluorescence 2 ug/ml, Immunocytochemistry 2 ug/ml |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18326285
|
Novus Biologicals
NBP3-15113-25UL |
25 μg |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18339125
|
Novus Biologicals
NBP3-15113-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APE Monoclonal antibody specifically detects APE in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, ImmunocytochemistrySpecifications
| APE | |
| Western Blot, ELISA 1:100-1:2000, Immunofluorescence 2 ug/ml, Immunocytochemistry 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AP endonuclease 1, APE, APE1, APE-1, APEAPEX nuclease (multifunctional DNA repair enzyme), APENAP endonuclease class I, APEX deoxyribonuclease (apurinic or apyrimidinic), APEX nuclease, APEX nuclease (multifunctional DNA repair enzyme) 1, Apurinic-apyrimidinic endonuclease 1, APXAP lyase, DNA-(apurinic or apyrimidinic site) lyase, EC 3.1, EC 4.2.99.18, HAP1apurinic/apyrimidinic (abasic) endonuclease, multifunctional DNA repair enzyme, protein REF-1, redox factor 1, Redox factor-1, REF-1, REF1 apurinic/apyrimidinic exonuclease | |
| A synthetic peptide of human APE (Uniprot#: P27695) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S07-5A9 | |
| Western Blot, ELISA, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 328 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title