missing translation for 'onlineSavingsMsg'
Learn More

APD597, MedChemExpress

Product Code. 16442727
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442727

Brand: Medchem Express HY15566

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress APD597 is a GPR119 agonist intended for the treatment of type 2 diabetes, with EC50 of 46 nM for hGPR119.

APD597 is a GPR119 agonist intended for the treatment of type 2 diabetes, with EC50 of 46 nM for hGPR119.

Specifications

Chemical Name or Material APD597
CAS 897732-93-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H29N5O6S
Quantity 50 mg
Synonym JNJ-38431055
Solubility Information DMSO : ≥ 19 mg/mL (39.62 mM)
SMILES O=C(OC(C)C)N(CC1)CCC1OC2=C(OC)C(NC3=CC=C(S(=O)(C)=O)N=C3C)=NC=N2
Molecular Weight (g/mol) 479.55
Formula Weight 479.55
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.