missing translation for 'onlineSavingsMsg'
Learn More

APD597, MedChemExpress

Product Code. 16469197
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469197 5 mg 5mg
16442727 50 mg 50mg
16426638 10 mg 10mg
16416816 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16469197 Supplier Medchem Express Supplier No. HY15566

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress APD597 is a GPR119 agonist intended for the treatment of type 2 diabetes, with EC50 of 46 nM for hGPR119.

APD597 is a GPR119 agonist intended for the treatment of type 2 diabetes, with EC50 of 46 nM for hGPR119.

Specifications

Chemical Name or Material APD597
CAS 897732-93-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H29N5O6S
Quantity 5 mg
Synonym JNJ-38431055
Solubility Information DMSO : ≥ 19 mg/mL (39.62 mM)
SMILES O=C(OC(C)C)N(CC1)CCC1OC2=C(OC)C(NC3=CC=C(S(=O)(C)=O)N=C3C)=NC=N2
Molecular Weight (g/mol) 479.55
Formula Weight 479.55
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.