missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542034
This item is not returnable.
View return policy
Description
Peptide sequence: ALSLDPNDQK SLEGMQKMEK EESPTDATQE EDVDDMEGSG EEGDLEGSDS Sequence homology: Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC7 is a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. The individual APC/C components mRNA and protein levels are expressed at approximately the same levels in most tissues and cell lines, suggesting that they perform their functions as part of a complex. APC7 is required for proper protein ubiquitination function of APC/C and for the interaction of APC/C with various transcription coactivators.
Specifications
| APC7 | |
| Polyclonal | |
| Unconjugated | |
| ANAPC7 | |
| Anapc7; anaphase promoting complex subunit 7; anaphase-promoting complex subunit 7; APC7; AW545589; Cyclosome subunit 7; prediabetic NOD sera-reactive autoantigen; si:ch211-127h20.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51434 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UJX3 | |
| ANAPC7 | |
| Synthetic peptide directed towards the C-terminal of human ANAPC7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction