missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APC6 Recombinant Rabbit Monoclonal Antibody (23GB3455), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553356
This item is not returnable.
View return policy
Description
The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
Specifications
| APC6 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Cdc16 | |
| 2700071J12Rik; 2810431D22Rik; ANAPC6; Anaphase-promoting complex subunit 6; anaphase-promoting complex, subunit 6; APC6; Cdc16; CDC16 cell division cycle 16; CDC16 cell division cycle 16 homolog; CDC16 homolog; CDC16Hs; cell division cycle 16; cell division cycle 16 homolog; cell division cycle protein 16 homolog; CUT9; Cyclosome subunit 6; RP11-569D9.4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 290875, 69957, 8881 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3455 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13042, Q8R349 | |
| Cdc16 | |
| A synthetic peptide of human Apc6/CDC16 (500-620AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction