missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APC6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12859092
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12859092 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12859092 Supplier Invitrogen™ Supplier No. PA520947

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human liver tissue lysate. PA5-20947 can be used with blocking peptide PEP-1061.

The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC6
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Cdc16
Gene Accession No. Q13042, Q8R349
Gene Alias 2700071J12Rik; 2810431D22Rik; ANAPC6; Anaphase-promoting complex subunit 6; anaphase-promoting complex, subunit 6; APC6; Cdc16; CDC16 cell division cycle 16; CDC16 cell division cycle 16 homolog; CDC16 homolog; CDC16Hs; cell division cycle 16; cell division cycle 16 homolog; cell division cycle protein 16 homolog; CUT9; Cyclosome subunit 6; RP11-569D9.4
Gene Symbols Cdc16
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human APC6.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69957, 8881
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.