missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APC6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583633
This item is not returnable.
View return policy
Description
Immunogen sequence: KDKLKCYDFD VHTMKTLKNI ISPPWDFREF EVEKQTAEET GLTPLETSRK TPDSRPSLEE TFEIEMNESD MMLETSMSDH ST Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 89%.
The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
Specifications
| APC6 | |
| Polyclonal | |
| Unconjugated | |
| Cdc16 | |
| 2700071J12Rik; 2810431D22Rik; ANAPC6; Anaphase-promoting complex subunit 6; anaphase-promoting complex, subunit 6; APC6; Cdc16; CDC16 cell division cycle 16; CDC16 cell division cycle 16 homolog; CDC16 homolog; CDC16Hs; cell division cycle 16; cell division cycle 16 homolog; cell division cycle protein 16 homolog; CUT9; Cyclosome subunit 6; RP11-569D9.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8881 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13042 | |
| Cdc16 | |
| Recombinant protein corresponding to Human CDC16. Recombinant protein control fragment (Product #RP-95122). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction