missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APC5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16330435
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16330435 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16330435 Supplier Invitrogen™ Supplier No. PA583478

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ADGAILDKGR AMFLVAKCQV ASAASYDQPK KAEALEAAIE NLNEAKNYFA KVDCKERIRD VVYFQARLYH TLGKTQERNR CAMLFRQLHQ ELPS Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 89%.

This gene encodes a tetratricopeptide repeat-containing component of the anaphase promoting complex/cyclosome, a large E3 ubiquitin ligase that controls cell cycle progression by targeting a number of cell cycle regulators such as B-type cyclins for 26S proteasome-mediated degradation through ubiquitination. The encoded protein is required for the proper ubiquitination function of APC/C and for the interaction of APC/C with transcription coactivators. It also interacts with polyA binding protein and represses internal ribosome entry site-mediated translation. Multiple transcript variants encoding different isoforms have been found for this gene. These differences cause translation initiation at a downstream AUG and result in a shorter protein, compared to isoform a.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Anapc5
Gene Accession No. Q9UJX4
Gene Alias 2510006G12Rik; AA408751; AA536819; AA986414; ANAPC5; anaphase promoting complex subunit 5; anaphase-promoting complex subunit 5; Anpc5; APC5; cyclosome subunit 5; MNCb-2778
Gene Symbols Anapc5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANAPC5. Recombinant protein control fragment (Product #RP-104708).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51433
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.