missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc5 Antibody (APC5#4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50106FR
This item is not returnable.
View return policy
Description
Apc5 Monoclonal specifically detects Apc5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Apc5 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ANAPC5 | |
| GST-human APC5 | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair | |
| 51433 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| APC5#4 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| anaphase promoting complex subunit 5, anaphase-promoting complex subunit 5, APC5Cyclosome subunit 5 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction