missing translation for 'onlineSavingsMsg'
Learn More

Apc5 Antibody (APC5#4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£150.00 - £302.00

Specifications

Antigen Apc5
Clone APC5#4
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Applications Western Blot, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18689448
View Documents
Novus Biologicals
NBP2-50106-0.025ml
0.025 mL
£150.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18617917
View Documents
Novus Biologicals
NBP2-50106
0.1 mL
£302.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Apc5 Monoclonal antibody specifically detects Apc5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications

Specifications

Apc5
1 mg/mL
Western Blot, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Cell Cycle and Replication, DNA Repair
PBS
51433
IgG1
Protein A purified
APC5#4
Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Monoclonal
Purified
RUO
Human
anaphase promoting complex subunit 5, anaphase-promoting complex subunit 5, APC5Cyclosome subunit 5
GST-human Apc5
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.