missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APC13 Polyclonal Antibody

Product Code. 12859102
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12859102 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12859102 Supplier Invitrogen™ Supplier No. PA520956

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Jurkat cell lysate. PA5-20956 can be used with blocking peptide PEP-1070.

Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC13 is a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. The individual APC/C components mRNA and protein levels are expressed at approximately the same levels in most tissues and cell lines, suggesting that they perform their functions as part of a complex. APC13 promotes the stable association of APC3/Cdc27 and APC6/Cdc16 with the APC/C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC13
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ANAPC13
Gene Accession No. Q8R034, Q9BS18
Gene Alias 1810004D07Rik; ANAPC13; anaphase promoting complex subunit 13; anaphase-promoting complex subunit 13; APC13; cyclosome subunit 13; cylosome subunit 13; SWM1
Gene Symbols ANAPC13
Host Species Rabbit
Immunogen A 17 amino acid peptide near the center of human APC13.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25847, 685029, 69010
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.