missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APC11 Polyclonal Antibody

Product Code. 12839102
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12839102 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12839102 Supplier Invitrogen™ Supplier No. PA520955

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is NIH-3T3 cell lysate. PA5-20955 can be used with blocking peptide PEP-1069.

Together with the cullin protein ANAPC2, constitutes the catalytic component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. The APC/C complex acts by mediating ubiquitination and subsequent degradation of target proteins: it mainly mediates the formation of 'Lys-11'-linked polyubiquitin chains and, to a lower extent, the formation of 'Lys-48'- and 'Lys-63'-linked polyubiquitin chains. May recruit the E2 ubiquitin-conjugating enzymes to the complex. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ANAPC11
Gene Accession No. Q9CPX9, Q9NYG5
Gene Alias 1110011I19Rik; Anapc11; anaphase promoting complex subunit 11; anaphase promoting complex subunit 11 (yeast APC11 homolog); anaphase promoting complex subunit 11 homolog; anaphase-promoting complex subunit 11; APC11; APC11 anaphase promoting complex subunit 11 homolog; Apc11p; Cyclosome subunit 11; hepatocellular carcinoma-associated RING finger protein; HSPC214; R75218; RGD1561880
Gene Symbols ANAPC11
Host Species Rabbit
Immunogen An 18 amino acid peptide near the center of human APC11.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498030, 51529, 66156
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.