missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APC11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554543
This item is not returnable.
View return policy
Description
Immunogen sequence: SPWCLDHSCD LFGITDQVSA DGPRACRQGA RRRLPAGVGP VLPLLPHALH PQVAARTAGA AALPHVPPGM EVQGV Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 29%.
Together with the cullin protein ANAPC2, constitutes the catalytic component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. The APC/C complex acts by mediating ubiquitination and subsequent degradation of target proteins: it mainly mediates the formation of 'Lys-11'-linked polyubiquitin chains and, to a lower extent, the formation of 'Lys-48'- and 'Lys-63'-linked polyubiquitin chains. May recruit the E2 ubiquitin-conjugating enzymes to the complex. [UniProt].
Specifications
| APC11 | |
| Polyclonal | |
| Unconjugated | |
| ANAPC11 | |
| 1110011I19Rik; Anapc11; anaphase promoting complex subunit 11; anaphase promoting complex subunit 11 (yeast APC11 homolog); anaphase promoting complex subunit 11 homolog; anaphase-promoting complex subunit 11; APC11; APC11 anaphase promoting complex subunit 11 homolog; Apc11p; Cyclosome subunit 11; hepatocellular carcinoma-associated RING finger protein; HSPC214; R75218; RGD1561880 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51529 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NYG5 | |
| ANAPC11 | |
| Recombinant protein corresponding to Human APC11. Recombinant protein control fragment (Product #RP-93303). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction