missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc11 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76260CL1
This item is not returnable.
View return policy
Description
Apc11 Polyclonal antibody specifically detects Apc11 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Apc11 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| anaphase promoting complex subunit 11, anaphase promoting complex subunit 11 (yeast APC11 homolog), anaphase-promoting complex subunit 11, APC11 anaphase promoting complex subunit 11 homolog, APC11Hepatocellular carcinoma-associated RING finger protein, Apc11p, Cyclosome subunit 11, HSPC214, MGC882 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human APC11. The immunogen is located within amino acids 70 - 120 of APC11. Amino Acid Squence: SLMSREESSRSPDPTPPA | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 51529 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction