missing translation for 'onlineSavingsMsg'
Learn More

Apc1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-86985B

Product Code. 18552821

  • £467.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Apc1 Polyclonal antibody specifically detects Apc1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Apc1
Polyclonal
Western Blot
anaphase promoting complex subunit 1, anaphase-promoting complex 1 (meiotic checkpoint regulator), anaphase-promoting complex subunit 1, APC1Testis-specific gene 24 protein, Cyclosome subunit 1, Mitotic checkpoint regulator, TSG24MCPR
The immunogen recognized by this antibody maps to a region between residue 500 and 550 of human anaphase promoting complex subunit 1 using the numbering given in entry NP_073153.1 (GeneID 64682).
0.1 mL
Cell Biology, Cell Cycle and Replication, Phospho Specific, Ubiquitin Proteasome Pathway
64682
Store at 4C in the dark.
IgG
Western Blot
Biotin
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.