missing translation for 'onlineSavingsMsg'
Learn More

Apc1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18418263 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418263 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418263 Supplier Novus Biologicals Supplier No. NBP177375AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Apc1 Polyclonal antibody specifically detects Apc1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apc1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias anaphase promoting complex subunit 1, anaphase-promoting complex 1 (meiotic checkpoint regulator), anaphase-promoting complex subunit 1, APC1Testis-specific gene 24 protein, Cyclosome subunit 1, Mitotic checkpoint regulator, TSG24MCPR
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human APC1. The immunogen is located within amino acids 1810 - 1860 of APC1. Amino Acid Squence: SRSHQERLQNHPKRGLF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 64682
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.