missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC-PCI Complex Antibody (M36), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41380AF594
This item is not returnable.
View return policy
Description
APC-PCI Complex Monoclonal specifically detects APC-PCI Complex in Human samples. It is validated for ELISA.
Specifications
| APC-PCI Complex | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mixture of PCI in complex with PSA (approx. 80%) and PCI cleaved from such a complex (approx. 20%). | |
| 100 μL | |
| Antibody is specific for APC:PCI complex and cleaved PCI but has little reactivity towards native PCI. The dissociation constants (as determined by standard surface plasmon resonance technique) are: Kd= 4x10e-10 M for PCI in complex with APC and 2x10e-10 M for cleaved PCI. The value of Kd for native PCI is too low to measure, i.e. > 10e-5 M. | |
| Store at 4°C in the dark. |
| ELISA | |
| M36 | |
| ELISA | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction