missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC Mouse anti-Human, Clone: ALi 12-28, Novus Biologicals™
Mouse Monoclonal Antibody
£395.00
Specifications
| Antigen | APC |
|---|---|
| Clone | ALi 12-28 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
Description
APC Monoclonal specifically detects APC in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| APC | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway | |
| 10 mM PBS with 0.05% BSA | |
| 324 | |
| IgG1 κ | |
| Protein A or G purified |
| ALi 12-28 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse (Negative) | |
| adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC | |
| Recombinant fragment of human APC protein (around aa 1-433) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title