missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC Antibody (Ali 12-28), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-667CL1
This item is not returnable.
View return policy
Description
APC Monoclonal antibody specifically detects APC in Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| APC | |
| Monoclonal | |
| CoraFluor 1 | |
| adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC | |
| N-terminal APC-MBP fusion protein | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway | |
| 324 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Ali 12-28 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction