missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APBB3 Antibody (OTI5G3), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71585PECY7
This item is not returnable.
View return policy
Description
APBB3 Monoclonal specifically detects APBB3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| APBB3 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| APBB3 | |
| Full length recombinant protein of human APBB3 (NP_573419) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI5G3 | |
| Flow Cytometry | |
| amyloid beta (A4) precursor protein-binding, family B, member 3, amyloid precursor interacting protein, Fe65L2, FE65L2amyloid beta A4 precursor protein-binding family B member 3, FE65-like protein 2, MGC150555, MGC87674, Protein Fe65-like 2, SRA | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 10307.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction