missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APBB3 Antibody (OTI5G3), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71585AF488
This item is not returnable.
View return policy
Description
APBB3 Monoclonal specifically detects APBB3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| APBB3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| APBB3 | |
| Full length recombinant protein of human APBB3 (NP_573419) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI5G3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| amyloid beta (A4) precursor protein-binding, family B, member 3, amyloid precursor interacting protein, Fe65L2, FE65L2amyloid beta A4 precursor protein-binding family B member 3, FE65-like protein 2, MGC150555, MGC87674, Protein Fe65-like 2, SRA | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 10307.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction