missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APBB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12680-50ul
This item is not returnable.
View return policy
Description
APBB3 Polyclonal antibody specifically detects APBB3 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| APBB3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate | |
| amyloid beta (A4) precursor protein-binding, family B, member 3, amyloid precursor interacting protein, Fe65L2, FE65L2amyloid beta A4 precursor protein-binding family B member 3, FE65-like protein 2, MGC150555, MGC87674, Protein Fe65-like 2, SRA | |
| Produced in rabbits immunized with E. coli-derived Human APBB3 fragment | |
| 50 μg | |
| Signal Transduction | |
| 10307 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction