missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
APBA3/Mint-3 Polyclonal antibody specifically detects APBA3/Mint-3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Gene Alias | Adapter protein X11gamma, amyloid beta (A4) precursor protein-binding, family A, member 3, amyloid beta (A4) precursor protein-binding, family A, member 3 (X11-like 2), MGC:15815, mint3, Mint-3, Neuronal Munc18-1-interacting protein 3, Neuron-specific X11L2 protein, phosphotyrosine-binding/-interacting domain (PTB)-bearing protein, X11L2amyloid beta A4 precursor protein-binding family A member 3, X11-like 2 protein |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human APBA3/Mint-3 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Primary or Secondary | Primary |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?