missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APBA3/Mint-3 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06040CL1
This item is not returnable.
View return policy
Description
APBA3/Mint-3 Polyclonal antibody specifically detects APBA3/Mint-3 in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| APBA3/Mint-3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| Adapter protein X11gamma, amyloid beta (A4) precursor protein-binding, family A, member 3, amyloid beta (A4) precursor protein-binding, family A, member 3 (X11-like 2), MGC:15815, mint3, Mint-3, Neuronal Munc18-1-interacting protein 3, Neuron-specific X11L2 protein, phosphotyrosine-binding/-interacting domain (PTB)-bearing protein, X11L2amyloid beta A4 precursor protein-binding family A member 3, X11-like 2 protein | |
| Produced in rabbits immunized with E. coli-derived Human APBA3/Mint-3 fragment. | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones, Vision | |
| 9546 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction