missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APBA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | APBA1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274772
|
Novus Biologicals
NBP2-56155 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610219
|
Novus Biologicals
NBP2-56155-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APBA1 Polyclonal specifically detects APBA1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| APBA1 | |
| Polyclonal | |
| Rabbit | |
| Membrane Trafficking and Chaperones, Vision | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 320 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ALPNHLHFHSLEHEEAMNAAYSGYVYTHRLFHRGEDEPYSEPYADYGGLQEHVYEEIGDAPELDARDGLRLYEQERDE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| Adapter protein X11alpha, amyloid beta (A4) precursor protein-binding, family A, member 1, amyloid beta A4 precursor protein-binding family A member 1, D9S411Eadaptor protein X11alpha, LIN10, Mint-1, MINT1amyloid beta (A4) precursor protein-binding, family A, member 1 (X11), Neuronal Munc18-1-interacting protein 1, Neuron-specific X11 protein, phosphotyrosine-binding/-interacting domain (PTB)-bearing protein, UROP11, X11A, X11X11ALPHA | |
| APBA1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title