missing translation for 'onlineSavingsMsg'
Learn More

Apatite, naturally occurring mineral, grains, approximately 0.06-0.19in

Product Code. 11399118
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399118 5 g 5g
11319128 1 g 1g
11309128 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399118 Supplier Thermo Scientific Alfa Aesar Supplier No. 042535.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals | CAS: 1306-05-4 | Ca5FO12P3 | 504.298 g/mol

Apatite is one of a few minerals produced and used by biological micro-environmental systems. The primary use of apatite is in the manufacture of fertilizer, it is a source of phosphorus. It is occasionally used as a gemstone. Green and blue varieties in finely divided form, are pigments with excellent covering power.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-05-4
Molecular Formula Ca5FO12P3
Molecular Weight (g/mol) 504.298
MDL Number MFCD00285511
InChI Key VSIIXMUUUJUKCM-UHFFFAOYSA-D
Synonym fluorapatite, apatite, phosphate rock, unii-m4cm1h238j, phospate rock, fluorapatite ca5f po4 3
PubChem CID 10207414
IUPAC Name pentacalcium;fluoride;triphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[F-].[Ca+2].[Ca+2].[Ca+2].[Ca+2].[Ca+2]

Specifications

Color Colorless
Physical Form Chips
Quantity 5 g
Solubility Information Soluble in water and hydrochloric acid.
Appearance Colorless chips
Chemical Name or Material Apatite
compliance-icons
Product Identifier
  • Apatite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Fluorapatite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.