missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APAF1 Polyclonal Antibody
Description
In Western blot applications, this antibody detects a band at ~130kDa. A suggested positive control is human heart tissue lysate.
APAF1 gene encodes a cytoplasmic protein that initiates apoptosis. This protein contains several copies of the WD-40 domain, a caspase recruitment domain (CARD), and an ATPase domain (NB-ARC). Upon binding cytochrome c and dATP, this protein forms an oligomeric apoptosome. The apoptosome binds and cleaves caspase 9 preproprotein, releasing its mature, activated form. Activated caspase 9 stimulates the subsequent caspase cascade that commits the cell to apoptosis. Alternative splicing results in several transcript variants encoding different isoforms.
Specifications
Specifications
| Antigen | APAF1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | APAF1 |
| Gene Accession No. | O14727, O88879, Q9EPV5 |
| Gene Alias | 6230400I06Rik; APAF1; APAF-1; Apaf1l; apoptotic peptidase activating factor 1; apoptotic protease activating factor 1; apoptotic protease-activating factor 1; CED4; DKFZp781B1145; fog; forebrain overgrowth; KIAA0413; mKIAA0413 |
| Gene Symbols | APAF1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?