missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APAF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583190
This item is not returnable.
View return policy
Description
Immunogen sequence: PLVVSLIGAL LRDFPNRWEY YLKQLQNKQF KRIRKSSSYD YEALDEAMSI SVEMLREDIK DYYTDLSILQ KDVKVPTKVL CILWDMETEE VEDILQEFVN KSLLFCDR Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 94%.
APAF1 gene encodes a cytoplasmic protein that initiates apoptosis. This protein contains several copies of the WD-40 domain, a caspase recruitment domain (CARD), and an ATPase domain (NB-ARC). Upon binding cytochrome c and dATP, this protein forms an oligomeric apoptosome. The apoptosome binds and cleaves caspase 9 preproprotein, releasing its mature, activated form. Activated caspase 9 stimulates the subsequent caspase cascade that commits the cell to apoptosis. Alternative splicing results in several transcript variants encoding different isoforms.
Specifications
| APAF1 | |
| Polyclonal | |
| Unconjugated | |
| APAF1 | |
| 6230400I06Rik; APAF1; APAF-1; Apaf1l; apoptotic peptidase activating factor 1; apoptotic protease activating factor 1; apoptotic protease-activating factor 1; CED4; DKFZp781B1145; fog; forebrain overgrowth; KIAA0413; mKIAA0413 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 317 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14727 | |
| APAF1 | |
| Recombinant protein corresponding to Human APAF1. Recombinant protein control fragment (Product #RP-95285). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction