missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APAF-1 Antibody (18H2), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80094B
This item is not returnable.
View return policy
Description
APAF-1 Monoclonal antibody specifically detects APAF-1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| APAF-1 | |
| Monoclonal | |
| Biotin | |
| APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413 | |
| Recombinant mouse APAF-1 (aa 1-97) containing the N-terminal CARD domain. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors | |
| 317 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunofluorescence | |
| 18H2 | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction