missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apabetalone, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Apabetalone (RVX-208) is an inhibitor of BET transcriptional regulators with selectivity for the second bromodomain. The IC50s are 87 μM and 0.51 μM for BD1 and BD2, respectively.
Specifications
Specifications
| Chemical Name or Material | Apabetalone |
| CAS | 1044870-39-4 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H22N2O5 |
| Quantity | 10 mg |
| Synonym | RVX-208 RVX000222 |
| Solubility Information | DMSO : ≥ 33 mg/mL (89.09 mM) |
| SMILES | COC1=CC(OC)=C(C(NC(C2=CC(C)=C(OCCO)C(C)=C2)=N3)=O)C3=C1 |
| Molecular Weight (g/mol) | 370.4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction