missing translation for 'onlineSavingsMsg'
Learn More

Apabetalone, MedChemExpress

Product Code. 16497098
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497098 10 mg 10mg
16449647 5 mg 5mg
16403147 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497098 Supplier Medchem Express Supplier No. HY16652
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Apabetalone (RVX-208) is an inhibitor of BET transcriptional regulators with selectivity for the second bromodomain. The IC50s are 87 μM and 0.51 μM for BD1 and BD2, respectively.

Apabetalone (RVX-208) is an inhibitor of BET transcriptional regulators with selectivity for the second bromodomain. The IC50s are 87 μM and 0.51 μM for BD1 and BD2, respectively.

Specifications

Chemical Name or Material Apabetalone
CAS 1044870-39-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H22N2O5
Quantity 10 mg
Synonym RVX-208 RVX000222
Solubility Information DMSO : ≥ 33 mg/mL (89.09 mM)
SMILES COC1=CC(OC)=C(C(NC(C2=CC(C)=C(OCCO)C(C)=C2)=N3)=O)C3=C1
Molecular Weight (g/mol) 370.4
Formula Weight 370.4
Percent Purity 98.19%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.