missing translation for 'onlineSavingsMsg'
Learn More

Apabetalone, MedChemExpress

Product Code. 16449647
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16449647

Brand: Medchem Express HY16652

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Apabetalone (RVX-208) is an inhibitor of BET transcriptional regulators with selectivity for the second bromodomain. The IC50s are 87 μM and 0.51 μM for BD1 and BD2, respectively.

Apabetalone (RVX-208) is an inhibitor of BET transcriptional regulators with selectivity for the second bromodomain. The IC50s are 87 μM and 0.51 μM for BD1 and BD2, respectively.

Specifications

Chemical Name or Material Apabetalone
CAS 1044870-39-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H22N2O5
Quantity 5 mg
Synonym RVX-208 RVX000222
Solubility Information DMSO : ≥ 33 mg/mL (89.09 mM)
SMILES COC1=CC(OC)=C(C(NC(C2=CC(C)=C(OCCO)C(C)=C2)=N3)=O)C3=C1
Molecular Weight (g/mol) 370.4
Formula Weight 370.4
Percent Purity 98.19%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.