missing translation for 'onlineSavingsMsg'
Learn More

AP4M1 Antibody, Novus Biologicals™

Product Code. 18399218 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399218 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399218 Supplier Novus Biologicals Supplier No. H00009179D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AP4M1 Polyclonal antibody specifically detects AP4M1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP4M1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_004713.2
Gene Alias Adapter-related protein complex 4 mu-1 subunit, adaptor-related protein complex 4, mu 1 subunit, adaptor-related protein complex AP-4 mu4 subunit, AP-4 adapter complex mu subunit, AP-4 complex subunit mu-1, CPSQ3, Mu subunit of AP-4, mu4, MU-4, Mu4-adaptin, Mu-adaptin-related protein 2, mu-adaptin-related protein-2, MUARP2, MU-ARP2
Host Species Rabbit
Immunogen AP4M1 (NP_004713.2, 1 a.a. - 453 a.a.) full-length human protein. MISQFFILSSKGDPLIYKDFRGDSGGRDVAELFYRKLTGLPGDESPVVMHHHGRHFIHIRHSGLYLVVTTSENVSPFSLLELLSRLATLLGDYCGSLGEGTISRNVALVYELLDEVLDYGYVQTTSTEMLRNFIQTEAVVSKPFSLFDLSSVGLFGAETQQSKVAPSSAASRPVLSSRSDQSQKNEVFLDVVERLSVLIASNGSLLKVDVQGEIRLKSFLPSGSEMRIGLTEEFCVGKSELRGYGPGIRVDEVSFHSSVNLDEFESHRILRLQPPQGELTVMRYQLSDDLPSPLPFRLFPSVQWDRGSGRLQVYLKLRCDLLSKSQALNVRLHLPLPRGVVSLSQELSSPEQKAELAEGALRWDLPRVQGGSQLSGLFQMDVPGPPGPPSHGLSTSASPLGLGPASLSFELPRHTCSGLQVRFLRLAFRPCGNANPHKWVRHLSHSDAYVIRI
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9179
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.