missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AP3S1 Polyclonal Antibody

Description
A suggested positive control is mouse kidney tissue lysate. PA5-21161 can be used with blocking peptide PEP-1275.
Part of the AP-3 complex, an adapter-related complex which is not clathrin-associated. The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes.
Specifications
Specifications
| Antigen | AP3S1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | AP3S1 |
| Gene Accession No. | Q92572, Q9DCR2 |
| Gene Alias | adapter-related protein complex 3 subunit sigma-1; adaptor related protein complex 3 sigma 1 subunit; adaptor related protein complex 3, sigma 1 subunit; adaptor-related protein complex 3 subunit sigma-1; adaptor-related protein complex 3, sigma 1 subunit; AP-3 complex subunit sigma-1; AP-3 complex subunit sigma-3A; AP3S1; CLAPS3; clathrin adaptor complex AP3, sigma-3A subunit; clathrin-associated/assembly/adapter protein, small 3; Clathrin-associated/assembly/adaptor protein, small 3; clathrin-associated/assembly/adaptor protein, small 3 (22kD); Sigma3A; Sigma-3A-adaptin; Sigma3A-adaptin; Sigma-adaptin 3a |
| Gene Symbols | AP3S1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?