missing translation for 'onlineSavingsMsg'
Learn More

AP3S1 Antibody, Novus Biologicals™

Product Code. 18333548 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333548 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333548 Supplier Novus Biologicals Supplier No. H00001176B02P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

AP3S1 Polyclonal antibody specifically detects AP3S1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP3S1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_001275.1
Gene Alias Adapter-related protein complex 3 sigma-1 subunit, adaptor-related protein complex 3, sigma 1 subunit, AP-3 complex sigma-3A subunit, AP-3 complex subunit sigma-3A, CLAPS3AP-3 complex subunit sigma-1, Clathrin-associated/assembly/adapter protein, small 3, clathrin-associated/assembly/adaptor protein, small 3 (22kD), Sigma3A, sigma-3A-adaptin, Sigma3A-adaptin, Sigma-adaptin 3a
Host Species Mouse
Immunogen AP3S1 (NP_001275.1, 1 a.a. - 193 a.a.) full-length human protein. MIKAILIFNNHGKPRLSKFYQPYSEDTQQQIIRETFHLVSKRDENVCNFLEGGLLIGGSDNKLIYRHYATLYFVFCVDSSESELGILDLIQVFVETLDKCFENVCELDLIFHVDKVHNILAEMVMGGMVLETNMNEIVTQIDAQNKLEKSEAGLAGAPARAVSAVKNMNLPEIPRNINIGDISIKVPNLPSFK
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1176
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.